Learn More
Medchemexpress LLC (1R,2S)-2-[(3,5-dichlorophenyl)carbamoyl]cyclohexane-1-carboxylic acid | 1093757-42-6 | 99.1% | C14H15Cl2NO3 | 10MG
SDP
Catalog No. 5000259795
Encompass_Preferred
VU0155041 is a small-molecule positive allosteric modulator of metabotropic glutamate receptor 4 (mGluR4) used in preclinical neuroscience and pharmacology research. It has reported EC50 values of approximately 798 nM (human) and 693 nM (rat), a molecular weight of 316.18 Da, molecular formula C14H15Cl2NO3, and CAS 1093757-42-6. The compound is supplied at high purity for laboratory research use only.
  • Potent, selective positive allosteric modulator of mGluR4.
  • Demonstrated activity in human and rat mGluR4 assays.
  • High purity for reproducible experimental results.
  • Soluble in DMSO for assay preparation.
  • Available in small research pack sizes suitable for preclinical studies.
  • Supplied for laboratory research use only.
Catalog No. 50-002-59795 Supplier Medchemexpress LLC Supplier No. HY1441710MG
VU0155041 is a small-molecule positive allosteric modulator of metabotropic glutamate receptor 4 (mGluR4) used in preclinical neuroscience and pharmacology research. It has reported EC50 values of approximately 798 nM (human) and 693 nM (rat), a molecular weight of 316.18 Da, molecular formula C14H15Cl2NO3, and CAS 1093757-42-6. The compound is supplied at high purity for laboratory research use only.
  • Potent, selective positive allosteric modulator of mGluR4.
  • Demonstrated activity in human and rat mGluR4 assays.
  • High purity for reproducible experimental results.
  • Soluble in DMSO for assay preparation.
  • Available in small research pack sizes suitable for preclinical studies.
  • Supplied for laboratory research use only.