Learn More
Medchemexpress LLC ((1R,2S,4R)-4-((5-(1-(3-bromobenzyl)-1H-pyrazole-3-carbonyl)pyrimidin-4-yl)amino)-2-hydroxycyclopentyl)methyl sulfamate | 1644342-14-2 | 99.8% | 551.41 | 1 ML
SDP
Catalog No. 5000346386
Encompass_Preferred
ML-792 is a potent and selective inhibitor of SAE/SUMO1 and SAE/SUMO2 in enzymatic assays with IC50 values of 3 and 11 nM, respectively. It shows significantly less activity against NAE/NEDD8 and UAE/ubiquitin. This compound inhibits SAE and SUMO-pathway activities in HCT116 cells, and decreases cancer cell viability in various cancer cell lines. This compound is for research use only.
  • Potent and selective inhibitor of SAE/SUMO1 and SAE/SUMO2.
  • Inhibits SAE and SUMO-pathway activities in HCT116 cells.
  • Inhibits cell proliferation and decreases cancer cell viability in various cancer cell lines (MDA-MB-468, MDA-MB-231, HCT116, Colo-205, and A375).
Catalog No. 50-003-46386 Supplier Medchemexpress LLC Supplier No. HY108702100MG
ML-792 is a potent and selective inhibitor of SAE/SUMO1 and SAE/SUMO2 in enzymatic assays with IC50 values of 3 and 11 nM, respectively. It shows significantly less activity against NAE/NEDD8 and UAE/ubiquitin. This compound inhibits SAE and SUMO-pathway activities in HCT116 cells, and decreases cancer cell viability in various cancer cell lines. This compound is for research use only.
  • Potent and selective inhibitor of SAE/SUMO1 and SAE/SUMO2.
  • Inhibits SAE and SUMO-pathway activities in HCT116 cells.
  • Inhibits cell proliferation and decreases cancer cell viability in various cancer cell lines (MDA-MB-468, MDA-MB-231, HCT116, Colo-205, and A375).