Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2S,5R)-(-)-Menthyl (S)-p-Toluenesulfinate 98.0+%, TCI America™
SDP

Catalog No. M104425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M104425G 25 g
M10445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M104425G Supplier TCI America Supplier No. M104425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1517-82-4
Molecular Formula C17H26O2S
Molecular Weight (g/mol) 294.453
MDL Number MFCD00010192
InChI Key NQICGNSARVCSGJ-DSRNLFJRSA-N
Synonym 1r,2s,5r---menthyl s-p-toluenesulfinate, 4-methylbenzenesulfinic acid 1r,5r-2-isopropyl-5-methylcyclohexyl ester
PubChem CID 101348873
IUPAC Name [(1R,5R)-5-methyl-2-propan-2-ylcyclohexyl] 4-methylbenzenesulfinate
SMILES CC1CCC(C(C1)OS(=O)C2=CC=C(C=C2)C)C(C)C

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 25 g
Formula Weight 294.45
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2S,5R)-(-)-Menthyl (S)-p-Toluenesulfinate
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.