Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (1R,2S)-VU0155041 | 1093757-42-6 | 99.1% | 316.18 g/mol | C14H15Cl2NO3 | 5 MG
SDP

Catalog No. 5000221589
Encompass_Preferred

VU0155041 is a research-grade small molecule that functions as a potent, selective positive allosteric modulator (PAM) of metabotropic glutamate receptor 4 (mGluR4). It has reported sub-micromolar activity and is supplied as a high-purity solid suitable for in vitro pharmacology and preclinical studies.

  • Potent, selective positive allosteric modulator of mGluR4 (EC50 ≈ 798 nM human, 693 nM rat).
  • High purity: 99.1%.
  • Molecular weight: 316.18 g/mol.
  • Soluble in DMSO at 100 mg/mL.
  • Supplied as a 5 mg solid for laboratory use.
  • Applicable to preclinical research, including Parkinson's disease models.

Catalog No. 50-002-21589 Supplier Medchemexpress LLC Supplier No. HY144175MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VU0155041 is a research-grade small molecule that functions as a potent, selective positive allosteric modulator (PAM) of metabotropic glutamate receptor 4 (mGluR4). It has reported sub-micromolar activity and is supplied as a high-purity solid suitable for in vitro pharmacology and preclinical studies.

  • Potent, selective positive allosteric modulator of mGluR4 (EC50 ≈ 798 nM human, 693 nM rat).
  • High purity: 99.1%.
  • Molecular weight: 316.18 g/mol.
  • Soluble in DMSO at 100 mg/mL.
  • Supplied as a 5 mg solid for laboratory use.
  • Applicable to preclinical research, including Parkinson's disease models.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.