Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,3S)-(-)-3-(Boc-amino)cyclopentanecarboxylic acid, 95%, Thermo Scientific Chemicals

Catalog No. AAH52022MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52022MD 250 mg
AAH5202203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52022MD Supplier Thermo Scientific Chemicals Supplier No. H52022MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 161660-94-2 | Molecular Formula: C11H18NO4 | Molecular Weight: 228.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 161660-94-2
Molecular Formula C11H18NO4
Molecular Weight (g/mol) 228.27
MDL Number MFCD01320858,MFCD01320857
InChI Key RNJQBGXOSAQQDG-SFYZADRCSA-M
Synonym 1r,3s-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, --1r,3s-n-boc-3-aminocyclopentanecarboxylic acid, cis-3-boc-amino cyclopentanecarboxylic acid, 1r,3s-3-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1r,3s-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1r,3s-boc-3-aminocyclopentane-1-carboxylic acid, cis-3-boc-amino cyclopentane-1-carboxylic acid, --1r,3s-n-boc-3-aminocyclopentane carboxylic acid
PubChem CID 1512526
SMILES CC(C)(C)OC(=O)N[C@H]1CC[C@H](C1)C([O-])=O

Specifications

IUPAC Name (1R,3S)-3-{[(tert-butoxy)carbonyl]amino}cyclopentane-1-carboxylate
Quantity 250 mg
Formula Weight 229.28
Percent Purity 95%
Chemical Name or Material (1R,3S)-(-)-3-(Boc-amino)cyclopentanecarboxylic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.