Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,3S)-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic Acid 97.0+%, TCI America™
SDP

Catalog No. B35381G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B35381G Supplier TCI America Supplier No. B35381G
Add to Cart
Add to Cart

High-purity chemical suitable for laboratory research use.

Chemical Identifiers

CAS 222530-39-4
Molecular Formula C12H21NO4
Molecular Weight (g/mol) 243.303
MDL Number MFCD04973107
InChI Key JSGHMGKJNZTKGF-BDAKNGLRSA-N
Synonym (1R,3S)-3-(Boc-amino)cyclohexanecarboxylic Acid
PubChem CID 16218560
IUPAC Name (1R,3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1CCCC(C1)C(=O)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 243.30
Percent Purity ≥97.0% (GC,T)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material (1R,3S)-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.