Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (1R,8S,9S)-bicyclo[6.1.0]non-4-yn-9-ylmethyl 2,5-dioxopyrrolidin-1-yl carbonate. | 1426827-79-3 | MFCD19705416 | 99.5% | 291.30 g/mol | C15H17NO5 | 100 MG
SDP

Catalog No. 5000299394
Encompass_Preferred

Endo-BCN-NHS carbonate is a succinimidyl carbonate reagent that installs bicyclo[6.1.0]non-4-yn (BCN) handles onto primary amines, enabling strain-promoted azide-alkyne cycloaddition (SPAAC) for copper-free click crosslinking. It is used to functionalize proteins and elastin-like polymers for hydrogel formation and cell-encapsulation studies.

  • High purity (99.54%).
  • Molecular formula C15H17NO5, molecular weight 291.30 g/mol.
  • Reacts with primary amines to introduce BCN handles for SPAAC crosslinking.
  • Appearance: white to light yellow solid.
  • Recommended storage: powder at -20°C; in solvent: -80°C (6 months) or -20°C (1 month).
  • Available in multiple package sizes including 100 mg.

Catalog No. 50-002-99394 Supplier Medchemexpress LLC Supplier No. HYW035129100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Endo-BCN-NHS carbonate is a succinimidyl carbonate reagent that installs bicyclo[6.1.0]non-4-yn (BCN) handles onto primary amines, enabling strain-promoted azide-alkyne cycloaddition (SPAAC) for copper-free click crosslinking. It is used to functionalize proteins and elastin-like polymers for hydrogel formation and cell-encapsulation studies.

  • High purity (99.54%).
  • Molecular formula C15H17NO5, molecular weight 291.30 g/mol.
  • Reacts with primary amines to introduce BCN handles for SPAAC crosslinking.
  • Appearance: white to light yellow solid.
  • Recommended storage: powder at -20°C; in solvent: -80°C (6 months) or -20°C (1 month).
  • Available in multiple package sizes including 100 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.