Learn More
Cayman Chemical 1R 9SHydrastIn 10mg
SDP
Catalog No. 502987658
Encompass_Preferred
An inhibitor of dopamine biosynthesis (IC50 = 20.7 µM in PC12 cells); inhibits tyrosine hydroxylase activity; inhibits dopamine release in the presence of a high extracellular K+ level (IC50 = 66.5 µM); inhibits L-type and caffeine-activated store-operated Ca2+ channels; prevents Ca2+ leakage from intracellular stores
Catalog No. 50-298-7658 Supplier Cayman Chemical Supplier No. 2236310 MG
An inhibitor of dopamine biosynthesis (IC50 = 20.7 µM in PC12 cells); inhibits tyrosine hydroxylase activity; inhibits dopamine release in the presence of a high extracellular K+ level (IC50 = 66.5 µM); inhibits L-type and caffeine-activated store-operated Ca2+ channels; prevents Ca2+ leakage from intracellular stores