Learn More
Cayman Chemical (1R,9S)-(–)-β-Hydrastine, Purity: ≥98%, CAS: 118-08-1, 50 mg
SDP
Catalog No. 502987938
Encompass_Preferred
An inhibitor of dopamine biosynthesis (IC50 = 20.7 µM in PC12 cells); inhibits tyrosine hydroxylase activity; inhibits dopamine release in the presence of a high extracellular K+ level (IC50 = 66.5 µM); inhibits L-type and caffeine-activated store-operated Ca2+ channels; prevents Ca2+ leakage from intracellular stores
Catalog No. 50-298-7938 Supplier Cayman Chemical Supplier No. 2236350 MG
An inhibitor of dopamine biosynthesis (IC50 = 20.7 µM in PC12 cells); inhibits tyrosine hydroxylase activity; inhibits dopamine release in the presence of a high extracellular K+ level (IC50 = 66.5 µM); inhibits L-type and caffeine-activated store-operated Ca2+ channels; prevents Ca2+ leakage from intracellular stores