Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1R-cis-Permethrin | 54774-45-7 | 99.9% | 5 MG
SDP

Catalog No. 5000408630
Encompass_Preferred

1R-cis-Permethrin (also known as 1R-cis-NRDC-143) is identified as an insecticide and neurotoxin. Its mechanism of action involves affecting neuron membranes by prolonging sodium channel activation.

  • Classification: insecticide, neurotoxin
  • Mechanism of action: prolongs sodium channel activation in neuron membranes
  • Uses: research purposes (not for human use)
  • Purity: 99.89% (100.00% ee)

Catalog No. 50-004-08630 Supplier Medchemexpress LLC Supplier No. HYB0887C5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1R-cis-Permethrin (also known as 1R-cis-NRDC-143) is identified as an insecticide and neurotoxin. Its mechanism of action involves affecting neuron membranes by prolonging sodium channel activation.

  • Classification: insecticide, neurotoxin
  • Mechanism of action: prolongs sodium channel activation in neuron membranes
  • Uses: research purposes (not for human use)
  • Purity: 99.89% (100.00% ee)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.