Learn More
Medchemexpress LLC (1r)-deruxtecan | 2270986-87-1 | 99.3% | 1,034.05 g·mol⁻¹ | C52H56FN9O13 | 5 MG
SDP
Catalog No. 5000222557
Encompass_Preferred
(1R)-Deruxtecan is a drug-linker conjugate intended for use in antibody-drug conjugate (ADC) research. It is supplied as a small-scale research reagent with supporting analytical documentation and is not for human or clinical use.
  • Drug-linker conjugate suitable for ADC development.
  • High purity (~99.3%) as indicated by analytical testing.
  • Available in small research pack sizes (5 mg).
  • Soluble in DMSO for preparation of stock solutions.
  • Provided with COA, SDS, and supporting analytical data when available.
  • Intended for research use only, not for human use.
Catalog No. 50-002-22557 Supplier Medchemexpress LLC Supplier No. HY13631O5MG
(1R)-Deruxtecan is a drug-linker conjugate intended for use in antibody-drug conjugate (ADC) research. It is supplied as a small-scale research reagent with supporting analytical documentation and is not for human or clinical use.
  • Drug-linker conjugate suitable for ADC development.
  • High purity (~99.3%) as indicated by analytical testing.
  • Available in small research pack sizes (5 mg).
  • Soluble in DMSO for preparation of stock solutions.
  • Provided with COA, SDS, and supporting analytical data when available.
  • Intended for research use only, not for human use.