Learn More
Medchemexpress LLC (1R)-IDH889 | 1429179-08-7 | 98.8% | 436.48 g/mol | C23H25FN6O2 | 5 MG
SDP
Catalog No. 5000263469
Encompass_Preferred
(1R)-IDH889 is the R-enantiomer of IDH889 supplied for use as an experimental control in studies of mutant isocitrate dehydrogenase 1 (IDH1). It is provided as a solid with high listed purity and recommended cold-storage conditions for both powder and solution forms.
  • R-enantiomer suitable as an experimental control in IDH1 assays.
  • Useful for comparing activity against mutant and wild-type IDH1.
  • High listed purity supports biochemical and cellular studies.
  • Stable as a powder under recommended cold storage conditions.
  • Available in multiple small-scale quantities and DMSO solution formats.
Catalog No. 50-002-63469 Supplier Medchemexpress LLC Supplier No. HY112289B5MG
(1R)-IDH889 is the R-enantiomer of IDH889 supplied for use as an experimental control in studies of mutant isocitrate dehydrogenase 1 (IDH1). It is provided as a solid with high listed purity and recommended cold-storage conditions for both powder and solution forms.
  • R-enantiomer suitable as an experimental control in IDH1 assays.
  • Useful for comparing activity against mutant and wild-type IDH1.
  • High listed purity supports biochemical and cellular studies.
  • Stable as a powder under recommended cold storage conditions.
  • Available in multiple small-scale quantities and DMSO solution formats.