Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (1R)-(+)-Nopinone

Catalog No. 3279565G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-795-65G 5 g
32-795-61G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 32-795-65G Supplier Sigma Aldrich Supplier No. 3279565G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(1R)-(+)-Nopinone may be used in the preparation of a chiral annulated indene derivative, which can be a potentially useful chiral ligand for transition metal complexes in asymmetric transformations. It may also react with secondary amines in cyclohexane to form the corresponding enamines.

Specifications

CAS 38651-65-9
Density 0.981 g/mL at 25°C
Boiling Point 209°C
Molecular Formula C9H14O
Refractive Index n20/D 1.479
Linear Formula C9H14O
MDL Number MFCD08447116
Quantity 5 g
Synonym 6,6-Dimethylbicyclo[3.1.1]heptan-2-one
Molecular Weight (g/mol) 138.21 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.