Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (1R)-tenofovir amibufenamide | 1571076-15-7 | 98.1% | C22H31N6O5P | 10MG
SDP

Catalog No. 5000258491
Encompass_Preferred

(1R)-Tenofovir amibufenamide is an investigational antiviral prodrug of tenofovir provided for research use. It is active against hepatitis B virus and HIV pathways and is supplied as a solid reagent for in vitro and preclinical studies; not for human or clinical use. Chemical identifiers include CAS 1571076-15-7, molecular formula C22H31N6O5P, and molecular weight 490.49 g/mol.

  • High purity 98.13%.
  • Molecular weight 490.49 g/mol.
  • Molecular formula C22H31N6O5P.
  • Active against HBV and HIV in preclinical assays.
  • Provided in milligram-scale quantities suitable for research.
  • Intended for in vitro and preclinical research, not for human use.

Catalog No. 50-002-58491 Supplier Medchemexpress LLC Supplier No. HY137453B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(1R)-Tenofovir amibufenamide is an investigational antiviral prodrug of tenofovir provided for research use. It is active against hepatitis B virus and HIV pathways and is supplied as a solid reagent for in vitro and preclinical studies; not for human or clinical use. Chemical identifiers include CAS 1571076-15-7, molecular formula C22H31N6O5P, and molecular weight 490.49 g/mol.

  • High purity 98.13%.
  • Molecular weight 490.49 g/mol.
  • Molecular formula C22H31N6O5P.
  • Active against HBV and HIV in preclinical assays.
  • Provided in milligram-scale quantities suitable for research.
  • Intended for in vitro and preclinical research, not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.