Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (1S,2R)-1-Aminoindan-2-ol | 126456-43-7 | 149.19 | 100 G
SDP

Catalog No. 5000448983
Encompass_Preferred

(1S,2R)-1-Aminoindan-2-ol is a chiral resolving agent designed to efficiently resolve various racemic 2-arylalkanoic acids. It precisely controls the stereostructure of products in asymmetric syntheses and can also be used in the synthesis of HIV protease inhibitors.

  • Chiral resolving agent
  • Efficiently resolves racemic 2-arylalkanoic acids
  • Controls stereostructure in asymmetric syntheses
  • Suitable for HIV protease inhibitor synthesis
  • White to off-white solid appearance
  • Purity: 100.0%
  • Molecular weight: 149.19
  • Store at 4°C, protect from light, stored under nitrogen

Catalog No. 50-004-48983 Supplier Medchemexpress LLC Supplier No. HYW002530100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(1S,2R)-1-Aminoindan-2-ol is a chiral resolving agent designed to efficiently resolve various racemic 2-arylalkanoic acids. It precisely controls the stereostructure of products in asymmetric syntheses and can also be used in the synthesis of HIV protease inhibitors.

  • Chiral resolving agent
  • Efficiently resolves racemic 2-arylalkanoic acids
  • Controls stereostructure in asymmetric syntheses
  • Suitable for HIV protease inhibitor synthesis
  • White to off-white solid appearance
  • Purity: 100.0%
  • Molecular weight: 149.19
  • Store at 4°C, protect from light, stored under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.