Learn More
Medchemexpress LLC (1S,2R)-1-Aminoindan-2-ol | 126456-43-7 | 149.19 | 25 G
SDP
Catalog No. 5000449462
Encompass_Preferred
This product is a chiral resolving agent that effectively resolves various racemic 2-arylalkanoic acids and precisely controls the stereostructure of the resulting products in asymmetric syntheses. It can also be utilized in the synthesis of HIV protease inhibitors, and is used as a raw material for the production of bulk drugs and active pharmaceutical ingredients.
  • Efficiently resolves racemic 2-arylalkanoic acids
  • Precisely controls the stereostructure of products
  • High purity (99.96%)
  • Utilized in the synthesis of HIV protease inhibitors
  • Used as a raw material for the production of bulk drugs
  • Material produced during the synthesis of an active pharmaceutical ingredient (API) that requires further molecular change or processing to become an API
Catalog No. 50-004-49462 Supplier Medchemexpress LLC Supplier No. 25GHYW002530
This product is a chiral resolving agent that effectively resolves various racemic 2-arylalkanoic acids and precisely controls the stereostructure of the resulting products in asymmetric syntheses. It can also be utilized in the synthesis of HIV protease inhibitors, and is used as a raw material for the production of bulk drugs and active pharmaceutical ingredients.
  • Efficiently resolves racemic 2-arylalkanoic acids
  • Precisely controls the stereostructure of products
  • High purity (99.96%)
  • Utilized in the synthesis of HIV protease inhibitors
  • Used as a raw material for the production of bulk drugs
  • Material produced during the synthesis of an active pharmaceutical ingredient (API) that requires further molecular change or processing to become an API