Learn More
Medchemexpress LLC (1S,2R)-1-Aminoindan-2-ol | 126456-43-7 | 149.19 | 500 G
SDP
Catalog No. 5000445902
Encompass_Preferred
(1S,2R)-1-Aminoindan-2-ol is a chiral resolving agent that efficiently resolves various racemic 2-arylalkanoic acids and precisely controls the stereostructure of the products in asymmetric syntheses. It can also be used in the synthesis of HIV protease inhibitors.
  • Chiral resolving agent
  • Efficiently resolves various racemic 2-arylalkanoic acids
  • Precisely controls stereostructure of products in asymmetric syntheses
  • Can be used in the synthesis of HIV protease inhibitors
Catalog No. 50-004-45902 Supplier Medchemexpress LLC Supplier No. HYW002530500G
(1S,2R)-1-Aminoindan-2-ol is a chiral resolving agent that efficiently resolves various racemic 2-arylalkanoic acids and precisely controls the stereostructure of the products in asymmetric syntheses. It can also be used in the synthesis of HIV protease inhibitors.
  • Chiral resolving agent
  • Efficiently resolves various racemic 2-arylalkanoic acids
  • Precisely controls stereostructure of products in asymmetric syntheses
  • Can be used in the synthesis of HIV protease inhibitors