Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,2R)-(-)-10,2-Camphorsultam, 99%

Catalog No. AAA1589706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1589706 5 g
AAA1589714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1589706 Supplier Thermo Scientific Chemicals Supplier No. A1589706
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 94594-90-8 | C10H17NO2S | 215.311 g/mol

(1S,2R)-(-)-10,2-Camphorsultam is used in the asymmetric synthesis of (S)- and (R)-N-Fmoc-S-trityl-α-methylcysteine. It is used as proton source in the synthesis of chiral α,γ-substituted γ-butyrolactones. It is also employed as a chiral probe for the optical resolution by HPLC and X-ray crystallographic determination of the absolute stereochemistry of carboxylic acids. Also used to prepare N-acryloyl derivatives which are employed as dienophiles in asymmetric Diels-Alder reactions. and for other asymmetric transformations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1S,2R)-(-)-10,2-Camphorsultam is used in the asymmetric synthesis of (S)- and (R)-N-Fmoc-S-trityl-α-methylcysteine. It is used as proton source in the synthesis of chiral α,γ-substituted γ-butyrolactones. It is also employed as a chiral probe for the optical resolution by HPLC and X-ray crystallographic determination of the absolute stereochemistry of carboxylic acids. Also used to prepare N-acryloyl derivatives which are employed as dienophiles in asymmetric Diels-Alder reactions. and for other asymmetric transformations.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94594-90-8
Molecular Formula C10H17NO2S
Molecular Weight (g/mol) 215.311
MDL Number MFCD00066271
InChI Key DPJYJNYYDJOJNO-SZBHIRRCSA-N
Synonym 5r,7r-10,10-dimethyl-3??-thia-4-azatricyclo 5.2.1.0 1 ,? decane-3,3-dione, --10,2-camphorsultam;--exo-10,2-bornanesultam; 3as-3a, a,6, a,7a, a-hexahydro-8,8-dimethyl-3h-3a,6-methano-2,1-benzisothiazole-2,2-dioxide; 1s,5r-10,10-dimethyl-3-thia-4-azatricyclo 5.2.1.01,5 decane 3,3-dioxide
PubChem CID 54579826
SMILES CC1(C2CCC13CS(=O)(=O)NC3C2)C

Specifications

Melting Point 181°C to 186°C
Quantity 5 g
Beilstein 83811
Solubility Information Slightly soluble in water.
Optical Rotation −33° (c=1 in Ethanol)
Formula Weight 215.32
Percent Purity 99%
Chemical Name or Material (1S,2R)-(-)-10,2-Camphorsultam
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.