Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (1S,2R)-2-Amino-1,2-diphenylethanol | 23364-44-5 | 100.0% | C14H15NO | 10 G
SDP

Catalog No. 5000434122
Encompass_Preferred

(1S,2R)-2-Amino-1,2-diphenylethanol is a biochemical reagent suitable for various life science-related research applications as a biological material or organic compound. It also acts as a potent and selective inhibitor of Histone deacetylase-3.

  • Functions as a biochemical reagent
  • Can be used as a biological material or organic compound
  • Potent and selective inhibitor of Histone deacetylase-3
  • Appears as a white to off-white solid
  • High purity of 99.96%
  • Molecular weight of 213.28
  • Soluble in DMSO

Catalog No. 50-004-34122 Supplier Medchemexpress LLC Supplier No. HY7627410G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(1S,2R)-2-Amino-1,2-diphenylethanol is a biochemical reagent suitable for various life science-related research applications as a biological material or organic compound. It also acts as a potent and selective inhibitor of Histone deacetylase-3.

  • Functions as a biochemical reagent
  • Can be used as a biological material or organic compound
  • Potent and selective inhibitor of Histone deacetylase-3
  • Appears as a white to off-white solid
  • High purity of 99.96%
  • Molecular weight of 213.28
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.