Learn More
Medchemexpress LLC (1S,2R)-2-Amino-1,2-diphenylethanol | 23364-44-5 | 100.0% | C14H15NO | 25 G
SDP
Catalog No. 5000359048
Encompass_Preferred
(1S,2R)-2-Amino-1,2-diphenylethanol is a biochemical reagent suitable for life science-related research. It acts as a potent and selective inhibitor of Histone deacetylase-3.
  • Can be used as a biological material or organic compound
  • Potent and selective inhibitor of Histone deacetylase-3
  • Suitable for in vitro research
Catalog No. 50-003-59048 Supplier Medchemexpress LLC Supplier No. 25GHY76274
(1S,2R)-2-Amino-1,2-diphenylethanol is a biochemical reagent suitable for life science-related research. It acts as a potent and selective inhibitor of Histone deacetylase-3.
  • Can be used as a biological material or organic compound
  • Potent and selective inhibitor of Histone deacetylase-3
  • Suitable for in vitro research