Learn More
Medchemexpress LLC (1S,2R)-2-Amino-1,2-diphenylethanol | 23364-44-5 | >99.96% | C14H15NO | 100 G
SDP
Catalog No. 5000434168
Encompass_Preferred
(1S,2R)-2-Amino-1,2-diphenylethanol is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is intended for research use only and not for sale to patients.
  • (1S,2R)-(+)-2-Amino-1,2-diphenylethanol are potent and selective inhibitors of Histone deacetylase-3.
Catalog No. 50-004-34168 Supplier Medchemexpress LLC Supplier No. HY76274100G
(1S,2R)-2-Amino-1,2-diphenylethanol is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is intended for research use only and not for sale to patients.
  • (1S,2R)-(+)-2-Amino-1,2-diphenylethanol are potent and selective inhibitors of Histone deacetylase-3.