Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (1S,2R)-2-Amino-1,2-diphenylethanol | 23364-44-5 | MFCD00074959 | 99.96% | C14H15NO | 50 G
SDP

Catalog No. 5000434528
Encompass_Preferred

As a potent and selective inhibitor of Histone deacetylase-3, (1S,2R)-2-Amino-1,2-diphenylethanol serves as a biochemical reagent and organic compound for life science research. This product is strictly for research use and not for patient application.

  • Purity of 99.96%
  • Molecular weight of 213.28
  • Chemical formula: C14H15NO
  • CAS number: 23364-44-5
  • Appears as a solid, white to off-white
  • Store at 4°C, protected from light, and under nitrogen
  • For solvent storage: -80°C for 6 months, or -20°C for 1 month (protected from light, under nitrogen)
  • Solubility in DMSO: 50 mg/mL (234.43 mM), ultrasonic assistance may be required

Catalog No. 50-004-34528 Supplier Medchemexpress LLC Supplier No. HY7627450G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

As a potent and selective inhibitor of Histone deacetylase-3, (1S,2R)-2-Amino-1,2-diphenylethanol serves as a biochemical reagent and organic compound for life science research. This product is strictly for research use and not for patient application.

  • Purity of 99.96%
  • Molecular weight of 213.28
  • Chemical formula: C14H15NO
  • CAS number: 23364-44-5
  • Appears as a solid, white to off-white
  • Store at 4°C, protected from light, and under nitrogen
  • For solvent storage: -80°C for 6 months, or -20°C for 1 month (protected from light, under nitrogen)
  • Solubility in DMSO: 50 mg/mL (234.43 mM), ultrasonic assistance may be required

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.