Learn More
Medchemexpress LLC (1S,2R)-2-amino-3-fluoro-1-(4-((methyl-d3)sulfonyl)phenyl)propan-1-ol | 00-00-0 | 97.3% | 250.30 | C10H11D3FNO3S | 5 MG
SDP
Catalog No. 5000262635
Encompass_Preferred
ent-Florfenicol amine-d3 is a deuterium-labeled internal standard of florfenicol amine used as a stable isotope tracer for LC-MS/MS method development and quantitative residue analysis. It is provided in small, high-purity quantities for analytical laboratories measuring veterinary drug metabolites and environmental residues.
  • Deuterium-labeled analogue enabling accurate isotopic internal standardization.
  • High purity (97.3%) suitable for quantitative analysis.
  • Supplied in small, ready-to-use quantities for LC-MS/MS workflows.
  • Stable under recommended storage conditions for reproducible results.
  • Applicable to veterinary residue and environmental testing.
Catalog No. 50-002-62635 Supplier Medchemexpress LLC Supplier No. HY133695S15MG
ent-Florfenicol amine-d3 is a deuterium-labeled internal standard of florfenicol amine used as a stable isotope tracer for LC-MS/MS method development and quantitative residue analysis. It is provided in small, high-purity quantities for analytical laboratories measuring veterinary drug metabolites and environmental residues.
  • Deuterium-labeled analogue enabling accurate isotopic internal standardization.
  • High purity (97.3%) suitable for quantitative analysis.
  • Supplied in small, ready-to-use quantities for LC-MS/MS workflows.
  • Stable under recommended storage conditions for reproducible results.
  • Applicable to veterinary residue and environmental testing.