Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (1S,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol | 15356-60-2 | MFCD00062983 | 100.0% | 156.27 g/mol | C10H20O | 1 ML
SDP

Catalog No. 5000301865
Encompass_Preferred

Ready-to-use 10 mM solution of (+)-menthol in DMSO for laboratory research. (+)-Menthol (IUPAC: (1S,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol; CAS 15356-60-2) is a monoterpene alcohol used in biochemical and pharmacological studies and supplied at high purity for reproducible results.

  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • High purity (99.95%) for consistent experimental results.
  • Molecular formula C10H20O; molecular weight 156.27 g/mol.
  • Includes datasheet, certificate of analysis, and safety data sheet.
  • Recommended storage: -80°C (6 months) or -20°C (1 month) for stock solutions.

Catalog No. 50-003-01865 Supplier Medchemexpress LLC Supplier No. HYW01727710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ready-to-use 10 mM solution of (+)-menthol in DMSO for laboratory research. (+)-Menthol (IUPAC: (1S,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol; CAS 15356-60-2) is a monoterpene alcohol used in biochemical and pharmacological studies and supplied at high purity for reproducible results.

  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • High purity (99.95%) for consistent experimental results.
  • Molecular formula C10H20O; molecular weight 156.27 g/mol.
  • Includes datasheet, certificate of analysis, and safety data sheet.
  • Recommended storage: -80°C (6 months) or -20°C (1 month) for stock solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.