Learn More
Medchemexpress LLC (1S,2R)-Alicapistat | 2221010-57-5 | 98.0% | 433.50 | 1 ML
SDP
Catalog No. 5000346650
Encompass_Preferred
(1S,2R)-Alicapistat is an orally active, selective inhibitor of human calpains 1 and 2, showing potential for Alzheimer's disease treatment. It mitigates metabolic liability of carbonyl reduction and inhibits calpain 1 with an IC50 value of 395 nM.
  • Orally active selective inhibitor of human calpains 1 and 2
  • Potential application for Alzheimer's disease
  • Mitigates metabolic liability of carbonyl reduction
  • Inhibits calpain 1 with an IC50 value of 395 nM
  • Prevents Aβ oligomer-induced deficits in synaptic transmission
  • Displays efficacy in preventing NMDA-induced neurodegeneration and A-induced synaptic dysfunction
Catalog No. 50-003-46650 Supplier Medchemexpress LLC Supplier No. HY109001A1MG
(1S,2R)-Alicapistat is an orally active, selective inhibitor of human calpains 1 and 2, showing potential for Alzheimer's disease treatment. It mitigates metabolic liability of carbonyl reduction and inhibits calpain 1 with an IC50 value of 395 nM.
  • Orally active selective inhibitor of human calpains 1 and 2
  • Potential application for Alzheimer's disease
  • Mitigates metabolic liability of carbonyl reduction
  • Inhibits calpain 1 with an IC50 value of 395 nM
  • Prevents Aβ oligomer-induced deficits in synaptic transmission
  • Displays efficacy in preventing NMDA-induced neurodegeneration and A-induced synaptic dysfunction