Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,2R)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine 98.0+%, TCI America™
SDP

Catalog No. B34835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34835G 5 g
B34831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34835G Supplier TCI America Supplier No. B34835G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 365996-30-1
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.309
MDL Number MFCD09952105
InChI Key AKVIZYGPJIWKOS-BDAKNGLRSA-N
Synonym (1S,2R)-N1-Boc-1,2-cyclohexanediamine, (1S,2R)-2-Amino-1-(tert-butoxycarbonylamino)cyclohexane, (1S,2R)-2-Amino-1-(Boc-amino)cyclohexane
PubChem CID 1514389
IUPAC Name tert-butyl N-[(1S,2R)-2-aminocyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCCCC1N

Specifications

Color White
Quantity 5 g
Formula Weight 214.31
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material (1S,2R)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.