Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,2S)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine 97.0+%, TCI America™
SDP

Catalog No. B2317500MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2317500MG 500 mg
B2317100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2317500MG Supplier TCI America Supplier No. B2317500MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 186769-18-6
Molecular Formula C20H28N2
Molecular Weight (g/mol) 296.458
MDL Number MFCD06797064
InChI Key ILMRHFMYIXTNMC-PMACEKPBSA-N
Synonym (1S,2S)-1,2-Diamino-1,2-dimesitylethane, (1S,2S)-1,2-Dimesitylethylenediamine
PubChem CID 11289465
IUPAC Name (1S,2S)-1,2-bis(2,4,6-trimethylphenyl)ethane-1,2-diamine
SMILES CC1=CC(=C(C(=C1)C)C(C(C2=C(C=C(C=C2C)C)C)N)N)C

Specifications

Melting Point 85°C
Color White-Yellow
Quantity 500 mg
Formula Weight 296.46
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (1S,2S)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.