Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,2S)-1,2-Bis(2-hydroxyphenyl)ethylenediamine 97.0+%, TCI America™
SDP

Catalog No. B30141G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B30141G Supplier TCI America Supplier No. B30141G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 870991-68-7
Molecular Formula C14H16N2O2
Molecular Weight (g/mol) 244.29
MDL Number MFCD09751761
InChI Key MRNPLGLZBUDMRE-UHFFFAOYNA-N
Synonym (1S,2S)-1,2-Diamino-1,2-bis(2-hydroxyphenyl)ethane, 2,2′C-[(1S,2S)-1,2-Diaminoethylene]bisphenol
PubChem CID 16218375
IUPAC Name 2-[1,2-diamino-2-(2-hydroxyphenyl)ethyl]phenol
SMILES NC(C(N)C1=CC=CC=C1O)C1=CC=CC=C1O

Specifications

Melting Point 159°C
Quantity 1 g
Formula Weight 244.29
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (1S,2S)-1,2-Bis(2-hydroxyphenyl)ethylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.