Learn More
Medchemexpress LLC (1S,2S)-2-[(1S)-1-amino-1-carboxy-2-(9H-xanthen-9-yl)ethyl]cyclopropane-1-carboxylic acid | 201943-63-7 | >98.0% | 353.4 g/mol | C20H19NO5 | 10 MG
SDP
Catalog No. 5000298149
Encompass_Preferred
Racemic LY341495 is a metabotropic glutamate receptor (mGluR) antagonist used as a research tool in neuroscience and pharmacology. It inhibits multiple mGlu receptor subtypes and is supplied as a crystalline solid for experimental use.
  • Antagonist of metabotropic glutamate receptors, particularly potent at group II (mGlu2, mGlu3).
  • Reported IC50s: mGlu2 = 21 nM, mGlu3 = 14 nM; higher micromolar activity at mGlu1a, mGlu5a, and mGlu4.
  • High purity: ≥98%.
  • Chemical formula C20H19NO5; molecular weight ≈ 353.4 g/mol.
  • Supplied as a small-quantity research pack (10 mg) for laboratory use.
Catalog No. 50-002-98149 Supplier Medchemexpress LLC Supplier No. HY70059A10MG
Racemic LY341495 is a metabotropic glutamate receptor (mGluR) antagonist used as a research tool in neuroscience and pharmacology. It inhibits multiple mGlu receptor subtypes and is supplied as a crystalline solid for experimental use.
  • Antagonist of metabotropic glutamate receptors, particularly potent at group II (mGlu2, mGlu3).
  • Reported IC50s: mGlu2 = 21 nM, mGlu3 = 14 nM; higher micromolar activity at mGlu1a, mGlu5a, and mGlu4.
  • High purity: ≥98%.
  • Chemical formula C20H19NO5; molecular weight ≈ 353.4 g/mol.
  • Supplied as a small-quantity research pack (10 mg) for laboratory use.