Learn More
Medchemexpress LLC (1S,2S)-2-PCCA hydrochloride | 1609563-70-3 | 99.7% | 528.56 | C30H39Cl2N3O | 10 MG
SDP
Catalog No. 5000215567
Encompass_Preferred
(1S,2S)-2-PCCA hydrochloride is the less-active diastereomer of 2-PCCA, a small-molecule GPR88 agonist. It inhibits GPR88-mediated cAMP production in HEK293 cells (EC50 = 116 nM) and is supplied as a solid hydrochloride salt for research applications, with defined purity and storage recommendations.
Catalog No. 50-002-15567 Supplier Medchemexpress LLC Supplier No. HY100013B110MG
(1S,2S)-2-PCCA hydrochloride is the less-active diastereomer of 2-PCCA, a small-molecule GPR88 agonist. It inhibits GPR88-mediated cAMP production in HEK293 cells (EC50 = 116 nM) and is supplied as a solid hydrochloride salt for research applications, with defined purity and storage recommendations.