Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (1S,2S)-2-PCCA hydrochloride | 1609563-70-3 | 99.7% | 528.56 g/mol | C30H39Cl2N3O | 1 ML
SDP

Catalog No. 5000215568
Encompass_Preferred

(1S,2S)-2-PCCA hydrochloride is the less active diastereomer of 2-PCCA, a small-molecule agonist of the GPR88 receptor. Supplied as a 10 mM solution in DMSO for in vitro receptor and cell-based assays, it inhibits GPR88-mediated cAMP production with a reported EC50 of 116 nM in HEK293 cells. The compound has a molecular weight of 528.56 g/mol and CAS number 1609563-70-3.

  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • Reported activity: inhibits GPR88-mediated cAMP production (EC50 116 nM in HEK293 cells).
  • High purity (99.7%) by LCMS.
  • Molecular weight 528.56 g/mol; formula C30H39Cl2N3O.
  • Intended for in vitro research; consult the safety data sheet for handling and storage.

Catalog No. 50-002-15568 Supplier Medchemexpress LLC Supplier No. HY100013B110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(1S,2S)-2-PCCA hydrochloride is the less active diastereomer of 2-PCCA, a small-molecule agonist of the GPR88 receptor. Supplied as a 10 mM solution in DMSO for in vitro receptor and cell-based assays, it inhibits GPR88-mediated cAMP production with a reported EC50 of 116 nM in HEK293 cells. The compound has a molecular weight of 528.56 g/mol and CAS number 1609563-70-3.

  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • Reported activity: inhibits GPR88-mediated cAMP production (EC50 116 nM in HEK293 cells).
  • High purity (99.7%) by LCMS.
  • Molecular weight 528.56 g/mol; formula C30H39Cl2N3O.
  • Intended for in vitro research; consult the safety data sheet for handling and storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.