Learn More
Medchemexpress LLC (1S,2S,3R)-DT-061 | 1809427-20-0 | 99.8% | 520.52 | 1 ML
SDP
Catalog No. 5000439765
Encompass_Preferred
(1S,2S,3R)-DT-061 is an enantiomer of DT-061, an orally active activator of protein phosphatase 2A (PP2A). This product can be used as a negative control in the research of KRAS-mutant and MYC-driven lung cancer tumorigenesis.
  • Enantiomer of DT-061
  • Orally active activator of protein phosphatase 2A (PP2A)
  • Used as a negative control in KRAS-mutant and MYC-driven lung cancer tumorigenesis research
Catalog No. 50-004-39765 Supplier Medchemexpress LLC Supplier No. HY112929B10MM/1ML
Add to Cart
Add to Cart
(1S,2S,3R)-DT-061 is an enantiomer of DT-061, an orally active activator of protein phosphatase 2A (PP2A). This product can be used as a negative control in the research of KRAS-mutant and MYC-driven lung cancer tumorigenesis.
  • Enantiomer of DT-061
  • Orally active activator of protein phosphatase 2A (PP2A)
  • Used as a negative control in KRAS-mutant and MYC-driven lung cancer tumorigenesis research