Learn More
Medchemexpress LLC (1S,2S,3R)-DT-061 | 1809427-20-0 | 99.8% | 520.52 | 25 MG
SDP
Catalog No. 5000439278
Encompass_Preferred
(1S,2S,3R)-DT-061 is an enantiomer of DT-061 and an orally active activator of protein phosphatase 2A (PP2A). It can be utilized as a negative control in research pertaining to KRAS-mutant and MYC-driven lung cancer tumorigenesis.
  • Orally active activator of protein phosphatase 2A (PP2A).
  • Can be utilized as a negative control in research.
  • Potential application in therapy of KRAS-mutant and MYC-driven tumorigenesis.
Catalog No. 50-004-39278 Supplier Medchemexpress LLC Supplier No. 25MGHY112929B
Add to Cart
Add to Cart
(1S,2S,3R)-DT-061 is an enantiomer of DT-061 and an orally active activator of protein phosphatase 2A (PP2A). It can be utilized as a negative control in research pertaining to KRAS-mutant and MYC-driven lung cancer tumorigenesis.
  • Orally active activator of protein phosphatase 2A (PP2A).
  • Can be utilized as a negative control in research.
  • Potential application in therapy of KRAS-mutant and MYC-driven tumorigenesis.