Learn More
Medchemexpress LLC HY-112929B 5mg Medchemexpress, (1S,2S,3R)-DT-061 CAS:1809427-20-0 Purity:>98%
SDP
Catalog No. 501932446
Encompass_Preferred
Medchemexpress, HY-112929B 5mg (1S,2S,3R)-DT-061 CAS:1809427-20-0 (1S,2S,3R)-DT-061 is an enantiomer of DT-061. DT-061 is an orally bioavailable activator of protein phosphatase 2A (PP2A) and could be applied in the therapy of KRAS-mutant and MYC-driven tumorigenesis [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2446 Supplier Medchemexpress LLC Supplier No. HY112929B5MG
Add to Cart
Add to Cart
Medchemexpress, HY-112929B 5mg (1S,2S,3R)-DT-061 CAS:1809427-20-0 (1S,2S,3R)-DT-061 is an enantiomer of DT-061. DT-061 is an orally bioavailable activator of protein phosphatase 2A (PP2A) and could be applied in the therapy of KRAS-mutant and MYC-driven tumorigenesis [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.