Learn More
Strem, An Ascensus Company CAS# 167316-27-0. 10g. (1S,2S)-(+)-N-(4-toluenesulfonyl)-1,2-diphenylethylenediamine, 98% (S,S)-TsDPEN. MFCD03095684
SDP
Catalog No. 5090109140
Encompass
CAS# 167316-27-0. 10g. (1S,2S)-(+)-N-(4-toluenesulfonyl)-1,2-diphenylethylenediamine, 98% (S,S)-TsDPEN. MFCD03095684. Molecular Weight: 366.48. Molecular Formula: C21H22N2O2S. Color/form: white xtl. Strem# 07-2370. http://www.strem.com/catalog/v/07-2370/
Catalog No. 50-901-09140 Supplier Strem, An Ascensus Company Supplier No. 07237010G
CAS# 167316-27-0. 10g. (1S,2S)-(+)-N-(4-toluenesulfonyl)-1,2-diphenylethylenediamine, 98% (S,S)-TsDPEN. MFCD03095684. Molecular Weight: 366.48. Molecular Formula: C21H22N2O2S. Color/form: white xtl. Strem# 07-2370. http://www.strem.com/catalog/v/07-2370/