Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (1S)-(+)-3-Carene

Catalog No. 4416195G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-161-95G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 44-161-95G Supplier Sigma Aldrich Supplier No. 4416195G
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(1S)-(+)-3-Carene is a monoterpene.Chiral building block for the preparation of bicyclo[3.2.0]heptenes that are useful intermediates in the synthesis of (+)-lineatin and both enantiomers of capnellene. Has been converted to its corresponding allenyl allylic ether for studies of its thermal isomerization chemistry. Precursor to di(4-isocaranyl)borane, an asymmetric hydroboration reagent for olefins.

Specifications

CAS 70862-65-6
Melting Point 163°C to 166°C
Molecular Formula C4H9NO3 -+ HCl
Refractive Index n20/D 1.472
Linear Formula HOCH2CH(NH2)CO2CH3-+HCl
MDL Number MFCD00066121
Quantity 5 g
Synonym (+)-3-Carene; (1S)-3,7,7-Trimethylbicyclo[4.1.0]hept-3-ene
Molecular Weight (g/mol) 155.58 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.