Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,3R)-3-Aminocyclohexanecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. A2286200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2286200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2286200MG Supplier TCI America Supplier No. A2286200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity crystalline powder for laboratory research applications.

Chemical Identifiers

CAS 81131-40-0
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
MDL Number MFCD09753648
InChI Key CKTUXQBZPWBFDX-NTSWFWBYSA-N
PubChem CID 7005079
IUPAC Name (1S,3R)-3-aminocyclohexane-1-carboxylic acid
SMILES C1CC(CC(C1)N)C(=O)O

Specifications

Melting Point 240°C
Color White
Quantity 200 mg
Formula Weight 143.19
Percent Purity ≥98.0% (GC,T)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material (1S,3R)-3-Aminocyclohexanecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.