Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,3R)-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic Acid 97.0+%, TCI America™
SDP

Catalog No. B35391G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B35391G Supplier TCI America Supplier No. B35391G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Crystalline powder used for organic synthesis applications.

Chemical Identifiers

CAS 222530-34-9
Molecular Formula C12H21NO4
Molecular Weight (g/mol) 243.303
MDL Number MFCD06227743
InChI Key JSGHMGKJNZTKGF-DTWKUNHWSA-N
Synonym (1S,3R)-3-(Boc-amino)cyclohexanecarboxylic Acid
PubChem CID 2755995
IUPAC Name (1S,3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1CCCC(C1)C(=O)O

Specifications

Color White
Quantity 1 g
Formula Weight 243.30
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (1S,3R)-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.