Learn More
Cayman Chemical 1S 3S3AmInomethylcyclope 50mg
SDP
Catalog No. 502951469
Encompass_Preferred
A polyphenolic antioxidant that is abundant in many fruits vegetables plant bark and peels has anti-carcinogenic anti-mutagenic anti-inflammatory and organ-preserving properties blocks methylation of H3R17 by CARM1 without significantly altering histone acetylase or DNA methyltransferase activity
Catalog No. 50-295-1469 Supplier Cayman Chemical Supplier No. 1135850 MG
A polyphenolic antioxidant that is abundant in many fruits vegetables plant bark and peels has anti-carcinogenic anti-mutagenic anti-inflammatory and organ-preserving properties blocks methylation of H3R17 by CARM1 without significantly altering histone acetylase or DNA methyltransferase activity