Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,4S)-2,5-Diazabicyclo[2.2.1]heptane Dihydrobromide 98.0+%, TCI America™
SDP

Catalog No. D26581G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D26581G Supplier TCI America Supplier No. D26581G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 132747-20-7
Molecular Formula C5H12Br2N2
Molecular Weight (g/mol) 259.97
MDL Number MFCD08272741
InChI Key ISYQWKOXKGJREA-UHFFFAOYNA-N
Synonym 1s,4s-2,5-diazabicyclo 2.2.1 heptane dihydrobromide, 1s,4s-+-2,5-diazabicyclo 2.2.1 heptane dihydrobromide, 1s,4s-2,5-diazabicylo 2.2.1 heptane dihydrobromide, 2,5-diazabicyclo 2.2.1 heptane,hydrobromide 1:2, 1s,4s-2,5-diazabicyclo 2.2.1 heptane dihydrobrom, 1s,4s-2,5-diaza-bicyclo 2.2.1 heptane dihydrobromide, 1s,2s-2,5-diazabicyclo 2.2.1 heptane dihydrobromide
PubChem CID 14612497
IUPAC Name 2,5-diazabicyclo[2.2.1]heptane dihydrobromide
SMILES Br.Br.C1NC2CNC1C2

Specifications

Quantity 1 g
Formula Weight 259.97
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (1S,4S)-2,5-Diazabicyclo[2.2.1]heptane Dihydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.