Learn More
Medchemexpress LLC (1S,4S)-N-desmethyl sertraline hydrochloride | 675126-10-0 | >95.0% | 328.66 g/mol | C16H16Cl3N | 1 MG
SDP
Catalog No. 5000280466
Encompass_Preferred
(1S,4S)-N-Desmethyl Sertraline hydrochloride is the N-desmethyl metabolite of the antidepressant sertraline and acts as a selective serotonin uptake blocker. It is used primarily in research applications - including metabolic profiling, pharmacology, and analytical method development - and is supplied as the hydrochloride salt with characterized molecular weight and CAS identifier.
Catalog No. 50-002-80466 Supplier Medchemexpress LLC Supplier No. HYW5875421MG
(1S,4S)-N-Desmethyl Sertraline hydrochloride is the N-desmethyl metabolite of the antidepressant sertraline and acts as a selective serotonin uptake blocker. It is used primarily in research applications - including metabolic profiling, pharmacology, and analytical method development - and is supplied as the hydrochloride salt with characterized molecular weight and CAS identifier.