Learn More
Cayman Chemical (1S,4S)-N-desmethyl Sertraline (hydrochloride), Purity: ≥95%, CAS: 675126-10-0, 10 mg
SDP
Catalog No. 502972246
Encompass_Preferred
A metabolite of sertraline; inhibits 5-HT and DA uptake in the rat corpus striatum (IC50s = 0.45 and 3.8 µM, respectively) and NE uptake in the rat hypothalamus (IC50 = 4.6 µM); does not increase extracellular levels of 5-HT or decrease extracellular levels of 5-HIAA in the rat striatum; does not increase cell firing in the rat dorsal raphe at 1,000 µg/kg, i.v.
Catalog No. 50-297-2246 Supplier Cayman Chemical Supplier No. 1591610 MG
A metabolite of sertraline; inhibits 5-HT and DA uptake in the rat corpus striatum (IC50s = 0.45 and 3.8 µM, respectively) and NE uptake in the rat hypothalamus (IC50 = 4.6 µM); does not increase extracellular levels of 5-HT or decrease extracellular levels of 5-HIAA in the rat striatum; does not increase cell firing in the rat dorsal raphe at 1,000 µg/kg, i.v.