Learn More
Cayman Chemical (1S)-(−)-α-Pinene, Purity: ≥95%, CAS: 7785-26-4, 50 g
SDP
Catalog No. 502910397
Encompass_Preferred
A bicyclic monoterpene with diverse biological activities; inhibits IBV replication (IC50 = 0.98 mM) and exhibits a CC50 value of >10 mM in Vero cells; active against several Gram-positive and Gram-negative bacteria, as well as the plant pathogenic fungus F. culmorum, and inhibits C. jejuni quorum sensing in vitro; cytotoxic to HCT116 cells from 125-500 µg/ml; oral administration reduces the colonization of fluoroquinoline-sensitive, but not fluoroquinoline-resistant, C. jejuni in broiler chicken cecum
Catalog No. 50-291-0397 Supplier Cayman Chemical Supplier No. 3569350 G
A bicyclic monoterpene with diverse biological activities; inhibits IBV replication (IC50 = 0.98 mM) and exhibits a CC50 value of >10 mM in Vero cells; active against several Gram-positive and Gram-negative bacteria, as well as the plant pathogenic fungus F. culmorum, and inhibits C. jejuni quorum sensing in vitro; cytotoxic to HCT116 cells from 125-500 µg/ml; oral administration reduces the colonization of fluoroquinoline-sensitive, but not fluoroquinoline-resistant, C. jejuni in broiler chicken cecum