Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S)-(-)-Camphanic acid, 99%, Thermo Scientific™

Catalog No. AAL1609803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1609803 1 g
AAL1609806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1609803 Supplier Thermo Scientific Chemicals Supplier No. L1609803

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13429-83-9
Molecular Formula C10H13O4
Molecular Weight (g/mol) 197.21
MDL Number MFCD00044948,MFCD00044948,MFCD00044948
InChI Key KPWKPGFLZGMMFX-ZJUUUORDSA-M
Synonym 1s-4,7,7-trimethyl-3-oxo-2-oxabicyclo 2.2.1 heptane-1-carboxylic acid, 1s---camphanic acid, 1s---camphanicacid, s---1-camphanic acid
PubChem CID 12302326
IUPAC Name (4S)-1,7,7-trimethyl-2-oxo-3-oxabicyclo[2.2.1]heptane-4-carboxylic acid
SMILES CC1(C)[C@]2(C)CC[C@]1(OC2=O)C([O-])=O

Specifications

Melting Point 198°C to 202°C
Quantity 1 g
Beilstein 84570
Formula Weight 198.22
Percent Purity 99%
Chemical Name or Material (1S)-(-)-Camphanic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-552-6
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.