Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S)-(+)-Camphorquinone 97.0+%, TCI America™
SDP

Catalog No. C16605G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16605G 5 g
C16601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16605G Supplier TCI America Supplier No. C16605G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2767-84-2
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00064159
InChI Key VNQXSTWCDUXYEZ-LHLIQPBNSA-N
Synonym (1S)-(+)-2,3-Bornanedione
PubChem CID 12222796
IUPAC Name (1S,4S)-4,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
SMILES CC1(C2CCC1(C(=O)C2=O)C)C

Specifications

Melting Point 200°C
Color White-Yellow
Quantity 5 g
Formula Weight 166.22
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (1S)-(+)-Camphorquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.