Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (1S)-(-)-Verbenone

Catalog No. 21825110G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-825-110G 10 g
21-825-150G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-825-110G Supplier Sigma Aldrich Supplier No. 21825110G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(1S)-(-)-Verbenone is one of the main components of the essential oil of Suregada zanzibariensis leaves. It can be prepared by the biotransformation of (-)-+a-pinene. (1S)-(-)-Verbenone acts as an antiaggregation pheromone towards the aggregation pheromone released by western pine beetle (WPB).(1S)-(-)-Verbenone can be used along with (1R)-(+)-nopinone for the stereoselective synthesis of chiral annulated indene derivatives via acid-catalyzed electrocyclic reaction. It can also be employed as a starting material in the preparation of verbenone derivatives containing a 4-styryl scaffold with potent anti-ischemic property.

Specifications

CAS 1196-01-6
Density 0.975 g/mL at 20°C
Boiling Point 227°C to 228°C
Molecular Formula C10H14O
Refractive Index n20/D 1.496
Linear Formula C10H14O
MDL Number MFCD00065445
Quantity 10 g
Synonym (1S,5S)-2-Pinen-4-one; (1S,5S)-4,6,6-Trimethylbicyclo[3.1.1]hept-3-en-2-one
Molecular Weight (g/mol) 150.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.