Learn More
Perkin Elmer US LLC 1-Slot Ceramic HybridXLT Quartz Torch
Catalog No. 502112244
Encompass_Preferred
PerkinElmer's HybridXLT torch is the latest addition to the family of PerkinElmer genuine sample introduction components. The new ceramic/quartz hybrid design provides the highest performance for your Optima ICP-OES 8x00 spectrometer. This new torch has an extended lifetime of up to 10x your current quartz torch, increasing instrument uptime and decreasing maintenance costs. This 1-slot torch is standard for aqueous solutions while the 3-slot HybridXLT torch (N0790249) is recommended for organic solvents. Features/Benefits: Extended Torch Lifetime - up to 10x improvement. Viewable plasma compared to a full ceramic design. Ideal for all sample types, except organic solvents. Increased instrument uptime. Lower cost per analysis. Lower cost than full ceramic torches.
Catalog No. 50-211-2244 Supplier Perkin Elmer US LLC Supplier No. N0780128
PerkinElmer's HybridXLT torch is the latest addition to the family of PerkinElmer genuine sample introduction components. The new ceramic/quartz hybrid design provides the highest performance for your Optima ICP-OES 8x00 spectrometer. This new torch has an extended lifetime of up to 10x your current quartz torch, increasing instrument uptime and decreasing maintenance costs. This 1-slot torch is standard for aqueous solutions while the 3-slot HybridXLT torch (N0790249) is recommended for organic solvents. Features/Benefits: Extended Torch Lifetime - up to 10x improvement. Viewable plasma compared to a full ceramic design. Ideal for all sample types, except organic solvents. Increased instrument uptime. Lower cost per analysis. Lower cost than full ceramic torches.