Learn More
Perkin Elmer US LLC 1-Slot Demountable Quartz Torch for Optima 8x00
Catalog No. 502112245
Encompass_Preferred
This demountable 1-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow and is the standard torch for aqueous solutions. Included in: Quick-Change Torch Module with Cyclonic Spray Chamber and Concentric Nebulizer (N0780607), Quick-Change Torch Module with Scott Spray Chamber and GemTip Cross-Flow II Nebulizer (N0780606), Scott/Cross-Flow II Sample Introduction Kit (N0780616), and Cyclonic/Concentric Sample Introduction Kit (N0780617) for Optima 8x00.
Catalog No. 50-211-2245 Supplier Perkin Elmer US LLC Supplier No. N0780130
This demountable 1-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow and is the standard torch for aqueous solutions. Included in: Quick-Change Torch Module with Cyclonic Spray Chamber and Concentric Nebulizer (N0780607), Quick-Change Torch Module with Scott Spray Chamber and GemTip Cross-Flow II Nebulizer (N0780606), Scott/Cross-Flow II Sample Introduction Kit (N0780616), and Cyclonic/Concentric Sample Introduction Kit (N0780617) for Optima 8x00.