Learn More
Perkin Elmer US LLC 1-Slot Demountable Quartz Torch for Optima 3x00 DV
Catalog No. 502113415
Encompass_Preferred
This demountable 1-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow and is the standard torch for aqueous solutions.
Catalog No. 50-211-3415 Supplier Perkin Elmer US LLC Supplier No. N0691662
This demountable 1-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow and is the standard torch for aqueous solutions.