Learn More
Enzo Life Sciences 1-Stearoyl-2-arachidonoyl-sn-glycerol (10mg). CAS: 65914-84-3
SDP
Supplier:  Enzo Life Sciences BMLDG1300010
Encompass_Preferred
Putative endogenous activator of protein kinase C produced via receptor-mediated inositol phospholipid hydrolysis. Purity: ≥95% (TLC). Solubility: Soluble in DMSO or 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-201-0512